Learn More
Brady™ Aluminum with Polyester Overlaminate Signs: Eating and Drinking Prohibited
Click to view available options
Diameter (Metric):
100 mm
200 mm
315 mm
Unit Size:
Each
Specifications
Specifications
| Alternate Part Number | PIC 224-DIA 100-B7525 |
| Diameter (Metric) | 100 mm |
| Shape | Round |