Learn More
OHAUS™ Aluminum Rod
OHAUS™ Aluminum Rod provides secure support when used with an OHAUS™ Lab Frame.
€ 6.25 - € 98.00
Product Code | Brand | Length (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Length (Metric) | Price | Quantity & Availability | |||||
15752319
|
OHAUS™
30392271 |
40 mm |
€ 6.25
Each |
Estimated Shipment: 22-05-2024
Log in to see stock. |
|||||
15762319
|
OHAUS™
30392272 |
50 mm |
€ 8.50
Each |
Estimated Shipment: 24-06-2024
Log in to see stock. |
|||||
15772319
|
OHAUS™
30392273 |
150 mm |
€ 10.75
Each |
Estimated Shipment: 24-06-2024
Log in to see stock. |
|||||
15782319
|
OHAUS™
30392274 |
300 mm |
€ 15.75
Each |
Estimated Shipment: 24-06-2024
Log in to see stock. |
|||||
15792319
|
OHAUS™
30392275 |
460 mm |
€ 17.00
Each |
Estimated Shipment: 24-06-2024
Log in to see stock. |
|||||
15702329
|
OHAUS™
30392276 |
610 mm |
€ 21.50
Each |
Estimated Shipment: 24-06-2024
Log in to see stock. |
|||||
15712329
|
OHAUS™
30392277 |
910 mm |
€ 24.00
Each |
Estimated Shipment: 24-06-2024
Log in to see stock. |
|||||
15722329
|
OHAUS™
30392278 |
1220 mm |
€ 37.00
Each |
Estimated Shipment: 24-06-2024
Log in to see stock. |
|||||
15732329
|
OHAUS™
30392279 |
1520 mm |
€ 50.40
Each |
Estimated Shipment: 24-06-2024
Log in to see stock. |
|||||
15742329
|
OHAUS™
30392280 |
1830 mm |
€ 47.50
Each |
Estimated Shipment: 24-06-2024
Log in to see stock. |
|||||
15752329
|
OHAUS™
30392281 |
2440 mm |
€ 60.50
Each |
Estimated Shipment: 24-06-2024
Log in to see stock. |
|||||
15772379
|
OHAUS™
30392333 |
3660 mm |
€ 98.00
Each |
Estimated Shipment: 24-06-2024
Log in to see stock. |
|||||
Description
- Corrosion-resistant lab-frames, rods and connectors are available in premium stainless steel, offering high chemical resistance and durability. Aluminum offers reliable strength at a lighter weight.
- Lab lifts offer convenient adjustable lifting platforms with exceptional strength and durability for holding small lab equipment such as glassware, hot plates and water baths at different heights.
- Simplify lab set up! Thirteen lab-frame kit options are available with four designed to fit in fume hoods. Build your own support system choosing from a wide selection of rods, bases and connectors.
- Dependable connectors secure rods and clamps to meet your requirements and five unique holders in a variety of sizes and materials accommodate different rods.
Specifications
Lab Frame Rod | |
Aluminum | |
13 mm |