Learn More
Bochem™ Aluminum Rods
Use Botech™ Aluminum Rods to support stand bases.
€ 12.50 - € 17.50
Product Code | Brand | Length (Metric) Rod | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Length (Metric) Rod | Price | Quantity & Availability | |||||
10400251
|
Bochem™
5143 |
1000 mm |
€ 17.50
Each |
Estimated Shipment: 03-05-2024
Log in to see stock. |
|||||
10676561
|
Bochem™
5140 |
500 mm |
€ 12.50
Each |
Estimated Shipment: 03-05-2024
Log in to see stock. |
|||||
10686561
|
Bochem™
5141 |
600 mm |
€ 13.40
Each |
Estimated Shipment: 13-05-2024
Log in to see stock. |
|||||
10248640
|
Bochem™
5132 |
750 mm |
€ 15.30
Each |
Estimated Shipment: 13-05-2024
Log in to see stock. |
|||||
Description
- Aluminum
- Without thread
- Sizes vary
- For stand base
- 12 D/O
Specifications
Aluminum |