Learn More
Korff™ Aluminum Round Bottom Evaporating Dishes
Eliminate the need for cleaning and storing of glass evaporating dishes. Korff™ Aluminum Round Bottom Evaporating Dishes are ideal for weighing, dispensing or moisture determinations.
Brand:  Korff™ 16605
Additional Details : Weight : 0.11900kg
Product Code. 10695792
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
  • Economical, lightweight aluminum dishes
  • Flared rim
  • Use as evaporating dishes or dust covers
Specifications
Specifications
Round
Aluminum
Evaporating Dish
13 mm
28 mL
Round
0.1 mm
64 mm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers