Learn More
Eppendorf™ Aluminum slide-out platform for I26 and I26R shakers
Product Code. 11622764
Click to view available options
For Use With (Equipment):
125 mL Erlenmeyer Flask Dedicated Platform
Unit Size:
Each
Product Code. 11622764
Brand: Eppendorf™ M13249905
Eppendorf shaker platforms are available in a wide variety of materials and sizes, perfect for virtually every requirement. Universal platforms can be adapted for thousands of combinations of clamps and other accessories. Dedicated platforms come complete with a single clamp size pre-installed.
Aluminum slide-out platform for I26 and I26R shakers, 76 × 46 cm, not interchangeable with other 76 x 46 cm platforms, 125 mL Erlenmeyer flask dedicated platform- Flask Capacity: 60
TRUSTED_SUSTAINABILITY
Specifications
Dimensions (L x W) 76 x 46 cm
Description Not interchangeable with other
For Use With (Equipment) 125 mL Erlenmeyer Flask Dedicated Platform