Learn More
VACUUBRAND™ Aluminum T-piece With Small KF Flanges
€ 53.00 - € 104.00
Product Code | Brand | Diameter (Metric) Inside | Size | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) Inside | Size | Price | Quantity & Availability | ||||
10039560
|
VACUUBRAND™
20669500 |
10/10/10 mm | 30 x 60 mm |
€ 53.00
Each |
Estimated Shipment: 19-06-2024
Log in to see stock. |
||||
10273371
|
VACUUBRAND™
20669504 |
16/16/16 mm | 40 x 80 mm |
€ 62.45
Each |
Estimated Shipment: 29-05-2024
Log in to see stock. |
||||
10450273
|
VACUUBRAND™
20669505 |
25/25/25 mm | 50 x 100 mm |
€ 74.55
Each |
Estimated Shipment: 27-05-2024
Log in to see stock. |
||||
10589911
|
VACUUBRAND™
20669506 |
40/40/40 mm | 65 x 130 mm |
€ 104.00
Each |
Estimated Shipment: 11-06-2024
Log in to see stock. |
||||
Specifications
Aluminum | |
1 Each | |
T-piece |