Learn More
WTW™ Aluminum Test Kits
Specifications
Specifications
| For Use With (Application) | For ground, surface, sea, waste water, drinking water, seepage water, process water |
| For Use With (Equipment) | For PhotoLab™ S12, PhotoLab™ Spectral, Photoflex photometers |
| Includes | Certificates |
| Range | 0.02 to 1.2 mg/L |
| Test Method | Photometric |
| Type | TOC Standard solution |