missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ALX1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA553876
This item is not returnable.
View return policy
Description
Immunogen sequence: QIQQAKSHFA ATYDISVLPR TDSYPQIQNN LWAGNASGGS VVTSCMLPRD TSSCMTPYSH SPRTDSSYTG FSNHQNQFSH VPLNNFFTDS LLTGATNGHA FETKPEFERR SSSIAVL Highest antigen sequence identity to the following orthologs: Mouse - 99%, Rat - 97%.
Transcriptional activator that acts at a palindromic recognition sequence to enhance the activity of the SV40 and TK promoters. Functions as a repressor with the prolactin promoter in vivo. May play a role in chondrocyte differentiation and may also influence cervix development.
Specifications
| ALX1 | |
| Polyclonal | |
| Unconjugated | |
| ALX1 | |
| AI314867; ALX homeobox 1; ALX homeobox protein 1; ALX1; C130005I02; Cart1; CART-1; CART1X; cartilage homeo protein 1; cartilage homeoprotein 1; cartilage paired-class homeoprotein 1; epididymis luminal protein 23; FND3; HEL23 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 8092 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q15699 | |
| ALX1 | |
| Recombinant protein corresponding to Human ALX1. Recombinant protein control fragment (Product #RP-100682). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction