missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ALX4 Antibody (OTI6B3) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72123
This item is not returnable.
View return policy
Description
ALX4 Monoclonal specifically detects ALX4 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
| ALX4 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1:2000, Immunohistochemistry 1:150, Immunocytochemistry/Immunofluorescence 1:100 | |
| ALX homeobox 4, FND2, homeobox protein aristaless-like 4, homeodomain transcription factor ALX4, KIAA1788 | |
| Mouse | |
| 44.1 kDa | |
| 100 μg | |
| Primary | |
| Reconstitute with PBS (pH 7.3). To use this carrier-free antibody for conjugation experiments, another round of desalting is highly recommended. | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG1 |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| OTI6B3 | |
| Unconjugated | |
| Lyophilized from PBS (pH 7.3) with 8% Trehalose with No Preservative | |
| ALX4 | |
| Full length human recombinant protein of human ALX4(NP_068745) produced in HEK293T cell. | |
| Affinity Purified | |
| Transcription Factors and Regulators | |
| 60529 | |
| Human, Mouse, Rat | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction