missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ALY Monoclonal Antibody (11G5), FITC

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA546554
This item is not returnable.
View return policy
Description
Western blot detects a band of approximately 32 kDa (predicted molecular weight: 27 kDa).
The protein encoded by this gene is a heat stable, nuclear protein and functions as a molecular chaperone. It is thought to regulate dimerization, DNA binding, and transcriptional activity of basic region-leucine zipper (bZIP) proteins.
Specifications
| ALY | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 0.09% sodium azide | |
| Q86V81 | |
| ALYREF | |
| Full length recombinant human ALY/Ref fusion protein. | |
| 50 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| ChIP Assay, ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Immunomicroscopy, Immunoprecipitation, Western Blot, Immunocytochemistry | |
| 11G5 | |
| FITC | |
| ALYREF | |
| ally of AML-1 and LEF-1; ALY; ALY/REF; Aly/REF export factor; Aly/REF export factor L homeolog; aly/REF export factor-A; alyref; alyref.L; alyref-a; BEF; bZIP enhancing factor; bZIP-enhancing factor BEF; hypothetical protein LOC537706; REF; Ref1; REF1-I; Refbp1; RNA and export factor binding protein 1; RNA and export factor-binding protein 1; Tcra enhancer-binding factor interacting protein 1; THO complex 4; THO complex subunit 4; THO complex subunit 4-A; tho4; Tho4-A; THOC4; thoc4-a; transcriptional coactivator Aly/REF; XELAEV_18043750mg; zgc:171753 | |
| Mouse | |
| Protein A | |
| RUO | |
| 10189 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. Store in the dark. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction