missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
This gene is a member of the DAP kinase-related apoptosis-inducing protein kinase family and encodes an autophosphorylated nuclear protein with a protein kinase domain. The protein has apoptosis-inducing activity.
Specifications
Specifications
| Antigen | ALY |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.27 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | ALYREF |
| Gene Accession No. | O08583, Q86V81 |
| Gene Alias | Ally of AML 1 and LEF 1, ALY, BEF, bZIP enhancing factor BEF, THO complex 4, THO complex subunit 4, Tho4, THOC4 |
| Gene Symbols | ALYREF |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?