missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AMACR Monoclonal Antibody (OTI3E10), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA808476
This item is not returnable.
View return policy
Description
This gene encodes a racemase. The encoded enzyme interconverts pristanoyl-CoA and C27-bile acylCoAs between their (R)- and (S)-stereoisomers. The conversion to the (S)-stereoisomers is necessary for degradation of these substrates by peroxisomal beta-oxidation. Encoded proteins from this locus localize to both mitochondria and peroxisomes. Mutations in this gene may be associated with adult-onset sensorimotor neuropathy, pigmentary retinopathy, and adrenomyeloneuropathy due to defects in bile acid synthesis. Alternatively spliced transcript variants have been described.Specifications
| AMACR | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9UHK6 | |
| AMACR | |
| Full length human recombit protein of human AMACR produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin) | |
| OTI3E10 | |
| Unconjugated | |
| AMACR | |
| AMACRD, CBAS4, P504S, RACE, RM | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 23600 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction