Learn More
Poulten & Graf GmbH Amber Glass Bottle with Screw Thread
Click to view available options
Capacity:
250 mL
500 mL
Bottle Surface:
Coated
Uncoated
Poulten & Graf GmbH Amber Glass Bottle with Screw Thread is a square bottles that can be used in a laboratory.
  • Square shape
  • 9.700 non-coated
  • 9.710 plastic coated
Bottle Surface Uncoated
Capacity 500 mL
Capacity (Metric) 500 mL
Description Amber glass bottle with screw thread
Form Square
Material Amber Glass
No. per Case 1
No. per Pack 1
Quantity 1
Sterility Non-sterile
Stopper Number 0
Transparency No
Type Bottle
Graduations No
Neck Finish Screw Thread
Pre-sterilized No
Certifications/Compliance EN ISO 4796-1
Autoclavable Autoclavable
Disposable No
Show More Show Less