missing translation for 'onlineSavingsMsg'
Learn More

behr Labor-Technik™ Clear Glass Bottle with Wide Neck

Product Code. 10444662
Click to view available options
Capacity (Metric):
250 mL
500 mL
Unit Size:
Pack of 10
This item is not returnable. View return policy

Product Code. 10444662

Brand: behr Labor-Technik™ B00218531

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Use these for storage, transport and sampling. behr Labor-Technik™ Clear Glass Bottles With Wide Necks are ideal for light-sensitive samples. Available in a wide range of sizes.

  • Clear glass bottle
  • Closure with PTFE insert
  • Available in packs of 10

Specifications

Capacity (Metric) 250 mL
Color Clear
For Use With (Application) For Storage, Transport and Sampling
Includes PTFE laminated screw cap
Material Glass
Quantity 10 Pk.
Mouth Wide
Product Type Storage Bottle
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.