missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences DURAN™ Amber Acid Storage Bottle Cap

Catalog Number 15251669
Click to view available options
Diameter (Metric):
48 mm
55 mm
66 mm
This item is not returnable. View return policy
This item is not returnable. View return policy

DURAN™ amber glass acid storage bottle cap.

Made of chemically resistant amber DURAN™ glass. With standard taper pennyhead stopper and additional ground glass cap. This provides an improved seal and protection against acid vapours. Ambered glass provide protection from Ultra-violet light.

  • Protection from UV light due to amber colouration
  • Ground DURAN™ glass stopper and additional DURAN™ glass cap provides protection against acid vapours
  • Pennyhead stopper with standard taper joint
  • Conical shoulders
  • Replacable glass cap
Diameter (Metric) 48 mm
For Use With (Equipment) For 100 mL bottle
Description Cap for 100 mL Bottle
For Use With (Application) Storage of photosensitive reagents
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'