missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Duran™ Amber Narrow Neck Reagent Bottle
Click to view available options
Capacity (Metric):
250 mL
Specifications
Diameter (Metric) | 72 mm |
Height (Metric) | 130 mm |
Product Type | Reagent Bottle |
Capacity (Metric) | 250 mL |
Includes | Standard Ground Joint |
Material | Soda Lime Glass |
Max. Temperature (Metric) | 100°C |
Quantity | 10Pack |
Stopper Number | 19/26 |
Certified | Yes |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'