Learn More
Invitrogen™ In Vivo GAPDH Positive Control siRNA
Ambion™ In Vivo GAPDH Positive Control siRNA
Merk:  Invitrogen™ 4457288
Overige gegevens : Gewicht : 0.01000kg
 Bekijk meerdere types van dit product via de
Produktcode. 15541491
  • € 253.00 / 1 stuk
Geschatte verzenddatum
bij leverancier 19-12-2018
Toevoegen aan winkelmandje
Beschrijving
Beschrijving
Invitrogen™ In Vivo siRNA Controls
  • Validated siRNA controls for optimizing siRNA experiments targeting coding and noncoding transcripts
  • Positive Control siRNAs functionally tested in several common cell lines
  • Negative Controls functionally proven to have minimal effects on cell proliferation and viability
  • Include Silencer™ Select modifications for enhanced specificity or Invitrogen™ In Vivo modifications for increased stability
  • For use in human, mouse, and rat cells
    What are Invitrogen™ In Vivo siRNAs?
    Invitrogen™ In Vivo siRNAs are designed using the proven Silencer™ Select algorithm and incorporate chemical modifications for superior serum stability with in vivo delivery. The added serum stability does not compromise the performance of the siRNAs. Invitrogen™ In Vivo siRNAs have been shown to be non-toxic in vivo (mouse) and non-immunogenic (cell-based assays). In cell-based assays, Invitrogen™ In Vivo siRNAs exhibit equivalent or better potency than Silencer™ Select siRNAs. The result is better confidence in your in vivo RNAi experiments, better knockdown when paired with an efficient delivery solution, and the assurance of limited non-specific effects.
    GAPDH Positive Control siRNAs
    Our extensively validated, positive control siRNA to human, mouse, and rat GAPDH serves multiple functions. First, it is an ideal “test” siRNA for those just beginning siRNA experiments, because it is validated to work in multiple cell lines. In addition, because it targets GAPDH mRNA, which is commonly used as an internal control, its effects are easy to assay, and thus provides an excellent tool to monitor siRNA transfection efficiency by real-time RT-PCR. The Invitrogen™ In Vivo GAPDH Positive Control siRNA shows increased serum and nuclease resistance compared with Silencer™ Select siRNAs without loss of potency or specificity and is recommended for in vivo applications.
    Quality Control
    Invitrogen™ synthesizes and purifies each Invitrogen™ In Vivo siRNA in state-of-the-art facilities to meet the highest quality standards. As part of our rigorous quality control procedures, each RNA oligonucleotide is analyzed by MALDI-TOF mass spectrometry, and analytical HPLC is used to monitor purity. To provide the utmost in quality, we also assess each annealed siRNA by gel electrophoresis to confirm that the strands anneal properly. The result is premium-quality siRNA that is purified and ready to use.
    In Vivo Ready Controls
    Our In Vivo Ready Control siRNAs are subjected to that extra level of purification and testing required for the introduction of siRNAs into animals. After HPLC purification and annealing, each siRNA is further purified utilizing a process that removes excess salt via a semi-permeable membrane. The result is highly pure siRNA with minimal salt content, suitable for in vivo applications. In vivo siRNAs are then sterile filtered, and tested for the presence of endotoxin. At concentrations of 50μM in presence of deionized water, in vivo Ready siRNAs contain <5.0 mM Na+, <0.06 mM K+, and <0.02 mM Mg2+.
  • Specificatie
    Specificatie
    Ambion™ In-vivo GAPDH Positive Control siRNA, 5nmol, Solid, Unconjugated Dye, siRNA RNAi Type, Gene Knockdown Positive Control Type, Used for RNAi, Transfection, GAPDH Target Gene, –20°C Storage Temperature, For Research Use Only
    RNAi, transfection
    Solid
    Ambion™ In-vivo GAPDH Positive Control siRNA
    –20°C
    Unconjugated
    Solid
    The siRNA (annealed) is shipped at ambient temperature as dried down oligonucleotides in a single tube, along with 1.75mL nuclease-free water for resuspension
    5nmol
    GAPDH