Learn More
Invitrogen™ Ambion™ RNase T1 (cloned) 1000U/μL
Endonuclease-free, exonuclease-free, protease-free
Brand: Invitrogen™ AM2280
Additional Details : Weight : 0.10500kg
Description
Pure preparation suitable for RNA mapping studies and other less demanding uses but priced similarly to less purified preparations.
- RNase T1 (E.C. 3.1.27.3) is isolated from over-expressing E. coli strain containing the cloned Aspergillus oryzae gene
- Endonuclease that specifically cleaves 3' of G residues
- Supplied in one tube containing 100,000U (1000U/μL
- RNase T1 is rigorously tested for contaminating nonspecific endonuclease, exonuclease, and protease activity
- Unit Definition: 100 units of RNase T1 is amount of enzyme that yields an increase in absorption at 260nm of 0.01428 units per min. at room temperature using 60μg/mL yeast total RNA as a substrate
RNA-Grade RNase T1 (Mfr. No. AM2283) is also available for use in RNA structure/function experiments.
DNA & RNA Purification & Analysis, Nuclease Protection Assays, Nucleic Acid Gel Electrophoresis & Blotting
Order Info
Shipping Conditions: Dry ice
Specifications
-20°C | |
100 μL |
For Research Use Only. Not for use in diagnostic procedures.