missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AMDHD1 Monoclonal Antibody (OTI4C1), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA809977S
This item is not returnable.
View return policy
Description
AMDHD1 Monoclonal antibody specifically detects AMDHD1 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western BlotSpecifications
| AMDHD1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9BZ95 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 54904 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG2a |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI4C1 | |
| Unconjugated | |
| Nsd3 | |
| 1300019J08Rik; Amdhd1; amidohydrolase domain containing 1; amidohydrolase domain-containing protein 1; HMFT1272; Probable imidazolonepropionase; RGD1308879 | |
| Human recombinant protein fragment corresponding to amino acids 427-566 of human WHSC1L1 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction