missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ AMELX Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5114845
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total AMELX.
This gene encodes a member of the amelogenin family of extracellular matrix proteins. Amelogenins are involved in biomineralization during tooth enamel development. Mutations in this gene cause X-linked amelogenesis imperfecta. Alternative splicing results in multiple transcript variants encoding different isoforms.
Specifications
| AMELX | |
| Polyclonal | |
| Unconjugated | |
| AMELX | |
| AI1E; AIH1; ALGN; Amel; amelogenesis imperfecta 1; amelogenin (amelogenesis imperfecta 1, X-linked); amelogenin (X chromosome, amelogenesis imperfecta 1); amelogenin X chromosome; amelogenin, X isoform; amelogenin, X-linked; Amelx; AMG; AMGL; AMGX; enamel matrix protein; leucine-rich amelogenin peptide; LRAP; M100888; Rgsc888 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 11704, 265 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide | |
| P63277, Q99217 | |
| AMELX | |
| A synthesized peptide derived from human AMELX(Accession Q99217), corresponding to amino acid residues T78-Q128. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Monkey | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction