missing translation for 'onlineSavingsMsg'
Learn More

Amenamevir, MedChemExpress

Product Code. 16436188
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
This item is not returnable. View return policy

Product Code. 16436188

Brand: Medchem Express HY14809

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Amenamevir is a helicase-primase inhibitor which has potent antiviral activity against HSVs with an EC50 of 14 ng/mL.

Amenamevir is a helicase-primase inhibitor which has potent antiviral activity against HSVs with an EC50 of 14 ng/mL.

Specifications

Chemical Name or Material Amenamevir
Physical Form Solid
CAS 841301-32-4
Color Light Yellow
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C24H26N4O5S
For Use With (Application) COVID-19-anti-virus
Synonym ASP2151
Solubility Information DMSO : ≥ 50 mg/mL (103.62 mM)
SMILES O=C(C(CC1)CCS1(=O)=O)N(C2=C(C)C=CC=C2C)CC(NC3=CC=C(C4=NOC=N4)C=C3)=O
Molecular Weight (g/mol) 482.55
Formula Weight 482.55
Percent Purity 99.89%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.