Learn More
Cytiva Amersham™ CyDye™ Value Packs (bis-Reactive NHS Ester)
Product Code. 11585035 Shop All Cytiva Products
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Product Type:
Cy2 bis-NHS Ester
Cy3 bis-NHS Ester
Cy5 Bis NHS Ester
Cy5 bis-NHS Ester
Cy5.5 Bis NHS Ester
Cy5.5 bis-NHS Ester
Cy7 Bis NHS Ester
Cy7 bis-NHS Ester
Unit Size:
10mg
50mg
5mg
Pack of 5
Product Code. 11585035
Brand: Cytiva PA12000
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Suitable for general labeling of amine residues
Order Info
Large range of bulk packs available for greatest economy. The packs contain fluors carrying two reactive groups on each dye molecule for general labeling of protein amine groups.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) Suitable for general labeling of Amine residues
Quantity 5 mg
Product Type Cy2 bis-NHS Ester
Kit Contents Foil packs, each containing 5, 10 or 50mg dried dye
Content And Storage 2°C to 8°C
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Skin sensitiser Category 1
  • Respiratory sensitiser Category 1