Description
- Greater concentration and more even distribution of positive charge than other charged nylon membranes
- Binding capacity for nucleic acids up to 600μg/cm2
- Efficient binding of small fragments
- Effective probe removal
- Suitable for reprobing
- Discs are also available
Specifications
Specifications
| Length (Metric) | 20 cm |
| Width (Metric) | 20 cm |
| For Use With (Equipment) | Radioactive probes |
| Binding Capacity | Up to 600μg/cm2 |
| Quantity | 10 Sheets |
| Format | Sheet |
| Material | Nylon, Highly Positively Charged |