Learn More
Cytiva Amersham™ Spacers for Vertical Gel Units
Shop All Cytiva Products
Click to view available options
Thickness (Metric):
0.07 cm
0.1 cm
0.15 cm
Width (Metric):
0.1 cm
2 cm
Unit Size:
Pack of 2
Specifications
Specifications
| For Use With (Equipment) | SE 400, SE 600 Vertical Gel Electrophoresis Unit |
| Type | Spacer |
| Thickness (English) | 0.02 in. |
| Thickness (Metric) | 0.07 cm |
| Length (Metric) | 16 cm |
| Width (Metric) | 2 cm |