Description
- Large surface area ensures very low breathing resistance
- New large exhalation valve which reduces heat build-up and improves wearer comfort
- color coded Alphamesh shell for ease of identification
- Adjustable headstrap allows for a secure and comfortable fit
- Metal-free - suitable for food industry applications
- Preformed nosebridge eliminates the need for regular adjustment
- Compatible with a wide range of safety eyewear
- Soft flexible mesh maintains its shape and will not collapse during use
- Conforms to EN 149: 2001 + A1: 2009 FFP2 NR
Specifications
Specifications
| Assigned Protection Factor (APF) | 10 |
| Certifications/Compliance | CE , EN 149:2000 + A1:2009 FFP2 NR |
| Color | Blue |
| Design Type | Cup/Molded |
| Disposable | Single-use |
| Filter Type | FFP2 |
| Material | Polypropylene |
| Product Type | General Purpose Particulate Respirator |
| Size | One Size |
| With Exhalation Valve | Yes |
| Show More |