missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AMFR/gp78 Antibody, PE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-49986PE
This item is not returnable.
View return policy
Description
AMFR/gp78 Polyclonal antibody specifically detects AMFR/gp78 in Human samples. It is validated for Western Blot, Immunoprecipitation
Specifications
| AMFR/gp78 | |
| Polyclonal | |
| Western Blot, Immunoprecipitation | |
| AMF receptor, AMF receptor, isoform 1, AMF receptor, isoform 2, autocrine motility factor receptor, Autocrine motility factor receptor, isoform 2, gp78, RING finger protein 45, RNF45EC 6.3.2.- | |
| The immunogen for this product maps to a region between residue 500 and 550 of human Autocrine Motility Factor Receptor using the numbering given in entry NP_001135.3 (GeneID 267). | |
| 0.1 mL | |
| Zinc Finger | |
| 267 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot, Immunoprecipitation | |
| PE | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction