missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AMG 487 (S-enantiomer), MedChemExpress
Description
AMG 487 S-enantiomer is the S enantiomer of AMG 487. AMG 487 is an antagonist of the chemokine receptor CXCR3.
Specifications
Specifications
| Chemical Name or Material | AMG 487 (S-enantiomer) |
| CAS | 473720-30-8 |
| Color | Light Yellow |
| Purity Grade Notes | Research |
| Molecular Formula | C32H28F3N5O4 |
| Quantity | 2 mg |
| Solubility Information | DMSO : 150 mg/mL (248.51 mM; Need ultrasonic) |
| SMILES | O=C(N([C@@H](C)C1=NC2=NC=CC=C2C(N1C3=CC=C(OCC)C=C3)=O)CC4=CC=CN=C4)CC5=CC=C(OC(F)(F)F)C=C5 |
| Molecular Weight (g/mol) | 603.59 |
| Formula Weight | 603.59 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?