missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ AMHR2 Recombinant Rabbit Monoclonal Antibody (8Z8X4)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA535722
This item is not returnable.
View return policy
Description
Immunogen sequence: PRAIRCLYSR CCFGIWNLTQ DRAQVEMQGC RDSDEPGCES LHCDPSPRAH PSPGSTLFTC SCGTDFCNAN YSHLPPPGSP GTPGSQGPQA APGESIWMAL V.
The AMH receptor (AMHR or AMHR2) is a serine/threonine kinase with a single transmembrane domain belonging to the family of type II receptors for TGF-beta-related proteins. Anti-Mullerian hormone (AMH) and its receptor are involved in the regression of Mullerian ducts in male fetuses. Male sex differentiation is mediated by 2 discrete hormones produced by the fetal testis. Testosterone, produced by Leydig cells, virilizes the external genitalia and promotes prostatic growth; anti-Mullerian hormone (AMH) results in regression of Mullerian ducts which would otherwise differentiate into the uterus and fallopian tubes.
Specifications
| AMHR2 | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q16671 | |
| AMHR2 | |
| A synthetic peptide corresponding to a sequence within amino acids 50-150 of human AMHR2 (NP_0654341). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| ELISA, Western Blot | |
| 8Z8X4 | |
| Unconjugated | |
| AMHR2 | |
| AMH type II receptor; AMHR; Amhr2; Anti-Muellerian hormone type II receptor; anti-Muellerian hormone type-2 receptor; anti-Mullerian hormone receptor type 11 SV1; anti-Mullerian hormone receptor type 2; anti-Mullerian hormone receptor type II; anti-Mullerian hormone receptor, type II; anti-Mullerian hormone type 2 receptor; anti-Mullerian hormone type 2 receptor delta 2; anti-Mullerian hormone type 2 receptor delta 9/10; C14; MIS type II receptor; Misiir; MISR2; MISRII; MRII; Muellerian inhibiting substance type II receptor; Mullerian inhibiting substance type II receptor | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 269 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction