missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AMICA/JAML Antibody (4E10), Alexa Fluor™ 488, Novus Biologicals™
Armenian Hamster Monoclonal Antibody
Brand: Novus Biologicals NBP1-43309AF488
This item is not returnable.
View return policy
Description
AMICA/JAML Monoclonal specifically detects AMICA/JAML in Mouse samples. It is validated for Flow Cytometry.
Specifications
| AMICA/JAML | |
| Monoclonal | |
| Alexa Fluor 488 | |
| adhesion molecule, interacts with CXADR antigen 1, AMICA, Gm638 | |
| Armenian Hamster | |
| Affinity Purified | |
| RUO | |
| 120425 | |
| Store at 4C in the dark. |
| Flow Cytometry | |
| 40000000000 | |
| Flow Cytometry | |
| AMICA1 | |
| The immunogen for this antibody was JAML. | |
| 0.1 mL | |
| Primary | |
| Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction