missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AMICA/JAML Antibody (4E10), PE/Atto594, Novus Biologicals™
Armenian Hamster Monoclonal Antibody
Brand: Novus Biologicals NBP1-43309PEATT594
This item is not returnable.
View return policy
Description
AMICA/JAML Monoclonal specifically detects AMICA/JAML in Mouse samples. It is validated for Flow Cytometry.
Specifications
| AMICA/JAML | |
| Monoclonal | |
| PE/ATTO 594 | |
| PBS with 0.05% Sodium Azide | |
| JAML | |
| Synthetic peptide corresponding to residues C(267) D G I L G D N F R P T Q(279) of mouse CARP. | |
| 0.1 mL | |
| 120425.0 | |
| Store at 4°C in the dark. Do not freeze. |
| Flow Cytometry | |
| 40000000000 | |
| Flow Cytometry | |
| adhesion molecule, interacts with CXADR antigen 1, AMICA, Gm638 | |
| Armenian Hamster | |
| Protein A or G purified | |
| Primary | |
| Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction