Learn More
Novus Biologicals™ AMID Antibody (Pab122), Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals™ NBP3-30820-25ug
 View more versions of this product
Product Code. 30067146
  • € 213.00 / 25µg
Description
AMID Monoclonal antibody specifically detects AMID in Hamster,Human,Monkey,Mouse,Rat samples. It is validated for Flow Cytometry,Immunoprecipitation,Western Blot,Immunohistochemistry (Paraffin),Immunohistochemistry (Frozen),Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY
Specifications
Specifications
AMID
Monoclonal
Unconjugated
Phosphate buffered saline
Mouse
Protein A or G purified
RUO
Primary
Hamster, Human, Monkey, Mouse, Rat
Purified
Flow Cytometry, Immunoprecipitation, Western Blot, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunocytochemistry/Immunofluorescence
Pab122
Western Blot : 5-10 μg/mL, Flow Cytometry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation : 2 μg/mg of protein lysate, Immunohistochemistry-Paraffin : 1-2 μg/mL, Immunohistochemistry-Frozen : 1-2 μg/mL
AMID, apoptosis-inducing factor (AIF)-like mitochondrion-associated inducer of death, apoptosis-inducing factor 2, Apoptosis-inducing factor homologous mitochondrion-associated inducer of death, apoptosis-inducing factor, mitochondrion-associated, 2, Apoptosis-inducing factor-like mitochondrion-associated inducer of death, DKFZp686L1298, FLJ14497, p53-responsive gene 3 protein, PRG3apoptosis-inducing factor (AIF)-homologous mitochondrion-associated inducer ofdeath, RP11-367H5.2
Hybridoma produced by the fusion of splenocytes from immunized with full length mouse AMID protein and mouse myeloma cells.
25 μg
Apoptosis, Cancer, Tumor Suppressors
84883
Store at -20°C. Avoid freeze/thaw cycles.
IgG2b