missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ AMID Polyclonal Antibody

Rabbit Polyclonal Antibody
€ 618.00 - € 620.00
Specifications
| Antigen | AMID |
|---|---|
| Concentration | 0.10 mg/mL |
| Content And Storage | Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
| Applications | Immunocytochemistry |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17128893
![]() |
Invitrogen™
PA5110968 |
100 μL |
€ 618.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17109073
![]() |
Invitrogen™
PA5111150 |
100 μL |
€ 620.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Immunogen sequence: NLVILCTGIK INSSAYRKAF ESRLASSGAL RVNEHLQVEG HSNVYAIGDC ADVRTPKMAY LAGLHANIAV ANIVNSVKQR PLQAYKPGAL TFLLSMGRND GVGQISGFYV GRLMVRLTKS RDLFVSTSWK TM Highest antigen sequence indentity to the following orthologs: Rat - 86%, Mouse - 86%.
The p53 tumor suppressor protein induces cell cycle arrest or apoptosis in response to cellular stresses. PRG3 (p53-responsive gene 3), is induced specifically under p53-dependent apoptotic conditions in human colon cancer cells. PRG3 has significant homology to bacterial oxidoreductases and the apoptosis-inducing factor, AIF. Unlike AIF, PRG3 localizes in the cytoplasm and its ectopic expression induces apoptosis. The PRG3 gene is thus a novel p53 target gene in a p53-dependent apoptosis pathway.Specifications
| AMID | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Polyclonal | |
| Liquid | |
| IgG | |
| Human | |
| AIFM2 | |
| 5430437E11Rik; AIFM2; Amid; apoptosis inducing factor, mitochondria associated 2; apoptosis-inducing factor (AIF)-homologous mitochondrion-associated inducer of death; apoptosis-inducing factor (AIF)-like mitochondrion-associated inducer of death; apoptosis-inducing factor 2; Apoptosis-inducing factor homologous mitochondrion-associated inducer of death; apoptosis-inducing factor, mitochondrion-associated 2; apoptosis-inducing factor, mitochondrion-associated, 2; apoptosis-inducing factor-like mitchondrion-associated inducer of death; apoptosis-inducing factor-like mitochondrion-associated inducer of death; D730001I10Rik; MBP2; MBPH; MGC126662; MGC141971; p53-responsive gene 3 protein; PRG3; RP11-367H5.2 | |
| AIFM2 | |
| Primary | |
| Antigen affinity chromatography |
| 0.10 mg/mL | |
| Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9BRQ8 | |
| 84883 | |
| Recombinant Protein corresonding to Human AMID. Recombinant protein control fragment (Product #RP-105802). | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title