missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Amifostine

Product Code. 18745536
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
This item is not returnable. View return policy

Product Code. 18745536

Brand: Tocris Bioscience™ 6655/100

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Cytoprotectant against chemotherapy and radiotherapy.

Amifostine is a cytoprotectant against chemotherapy and radiotherapy. Reduces mitochondrial pyruvate use in hepatocytes but not hepatoma cells in vitro. Also inhibits angiogenesis.

Active metabolite is also available, WR 1065 (Cat. No. 3356)

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-[(3-Aminopropyl)amino]ethanethiol dihydrogen phosphate
CAS 20537-88-6
Quantity 100 mg
Molecular Formula C5H15N2O3PS
Formula Weight 214.22
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.