missing translation for 'onlineSavingsMsg'
Learn More

Melford Amikacin

Product Code. 30018008
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 30018008

Brand: Melford A2053025.0

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Amikacin is an aminoglycoside antibiotic.

Amikacin is an aminoglycoside antibiotic. It is often utilized in microbiology and molecular biology laboratories and functions by inhibiting bacterial protein synthesis. Additionally, it binds to the 30S ribosomal subunit of bacteria, leading to the misreading of mRNA and ultimately preventing the synthesis of bacterial proteins. Due to its mechanism of action, amikacin is effective against a wide range of Gram-negative and some Gram-positive bacteria.

Commonly used as a selective agent in cell culture and molecular biology experiments. May be included in culture media to selectively grow cells that have been genetically modified to express resistance to this antibiotic. This is often achieved by introducing a plasmid or vector containing an amikacin resistance gene into the cells.

Specifications

Chemical Name or Material Amikacin
CAS 37517-28-5
Quantity 25 g
Molecular Formula C22H43N5O13
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.