missing translation for 'onlineSavingsMsg'
Learn More

Amino Albendazole, TRC

Product Code. 30316636
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30316636 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30316636 Supplier Toronto Research Chemicals Supplier No. TRCA580945500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 5-(Propylsulphanyl)-1H-benzimidazol-2-amine
CAS 80983-36-4
Molecular Formula C10 H13 N3 S
Quantity 500 mg
Synonym 1H-Benzimidazol-2-amine, 5-(propylthio)- (9CI), 6-(Propylthio)-1H-benzimidazol-2-amine, 5-(Propylthio)-1H-benzimidazol-2-amine
InChI Formula InChI=1S/C10H13N3S/c1-2-5-14-7-3-4-8-9(6-7)13-10(11)12-8/h3-4,6H,2,5H2,1H3,(H3,11,12,13)
SMILES CCCSc1ccc2[nH]c(N)nc2c1
IUPAC Name 5-propylsulfanyl-1H-benzimidazol-2-amine
Molecular Weight (g/mol) 207.3
Formula Weight 207.083
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.