missing translation for 'onlineSavingsMsg'
Learn More

Aminoacetonitrile Hydrochloride, TRC

Product Code. 30323330
Click to view available options
Quantity:
25 g
Unit Size:
25g
This item is not returnable. View return policy

Product Code. 30323330

Brand: Toronto Research Chemicals TRCA58005525G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Aminoacetonitrile Hydrochloride
CAS 6011-14-9
Molecular Formula C2H5ClN2
Quantity 25 g
Synonym 2-Amino-acetonitrile Hydrochloride, Amino-acetonitrile Monohydrochloride, Glycinonitrile Monohydrochloride, 2-Aminoacetonitrile Hydrochloride, Aminoacetonitrile Hydrochloride, Aminoacetonitrile Monohydrochloride, Glycinonitrile Hydrochloride, alpha-Aminoacetonitrile Hydrochloride
InChI Formula InChI=1S/C2H4N2.ClH/c3-1-2-4;/h1,3H2;1H
SMILES Cl.NCC#N
IUPAC Name 2-aminoacetonitrile;hydrochloride
Molecular Weight (g/mol) 92.53
Formula Weight 92.0141
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.