Learn More
Aminoguanidine bicarbonate, 98.5%, Thermo Scientific Chemicals
Catalog Number p-4510103
Click to view available options
Quantity:
250 g
1 kg
5 kg
Packaging:
Plastic Bottle
Plastic drum
2582-30-1
CH6N4·H2CO3
136.11
OTXHZHQQWQTQMW-UHFFFAOYSA-N
aminoguanidine bicarbonate, aminoguanidine hydrogen carbonate, aminoguanidinium bicarbonate, aminoguanidine hydrocarbonate, aminoguanidine carbonate 1:1, aminoguanidium hydrogen carbonate, n1-aminoguanidine carbonate 1:1, aminoguanidine hydrogencarbonate, aminoguanidinebicarbonate, hydrazinecarboximidamide carbonate
164944
2-aminoguanidine;carbonic acid
C(=NN)(N)N.C(=O)(O)O
2582-30-1
CH6N4·H2CO3
136.11
OTXHZHQQWQTQMW-UHFFFAOYSA-N
aminoguanidine bicarbonate, aminoguanidine hydrogen carbonate, aminoguanidinium bicarbonate, aminoguanidine hydrocarbonate, aminoguanidine carbonate 1:1, aminoguanidium hydrogen carbonate, n1-aminoguanidine carbonate 1:1, aminoguanidine hydrogencarbonate, aminoguanidinebicarbonate, hydrazinecarboximidamide carbonate
164944
2-aminoguanidine;carbonic acid
C(=NN)(N)N.C(=O)(O)O
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Melting Point 125°C
Color White
Ignition Residue 0.3% max.
Infrared Spectrum Authentic
Assay Percent Range 98.5%
Packaging Plastic Bottle
Linear Formula H2NNHC(=NH)NH2·H2CO3
Quantity 250 g
Beilstein 03,117
Solubility Information Solubility in water: <5g/L (20°C)
Formula Weight 136.11
Percent Purity 98.50%
Physical Form Crystalline Powder
Chemical Name or Material Aminoguanidine bicarbonate
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Aminoguanidine bicarbonate
Signal Word
  • Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Respiratory sensitiser Category 1
Hazard Statement
  • H411-Toxic to aquatic life with long lasting effects.
  • H317-May cause an allergic skin reaction.
Precautionary Statement
  • P273-Avoid release to the environment.