missing translation for 'onlineSavingsMsg'
Learn More

Aminomethyltransferase Antibody (OTI6F4), Alexa Fluor™ 594, Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals NBP2-70171AF594

Product Code. 18105623

  • € 751.00 / 0.10mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Aminomethyltransferase Monoclonal specifically detects Aminomethyltransferase in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Aminomethyltransferase
Monoclonal
Alexa Fluor 594
50 mM sodium borate with 0.05% sodium azide
AMT
Recombinant protein fragment corresponding to amino acids 146-403 of human Aminomethyltransferase (NP_000472) produced in E.coli.
0.1 mL
Primary
Human, Mouse, Rat
IgG1
Western Blot, Immunohistochemistry
OTI6F4
Western Blot, Immunohistochemistry
aminomethyltransferase, EC 2.1.2.10, GCE, GCSTaminomethyltransferase (glycine cleavage system protein T), glycine cleavage system protein T, NKHmitochondrial
Mouse
Affinity Purified
Neuroscience
275.0
Store at 4°C in the dark.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.