missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aminopeptidase A/ENPEP Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 227.00 - € 326.00
Specifications
| Antigen | Aminopeptidase A/ENPEP |
|---|---|
| Dilution | Immunocytochemistry/ Immunofluorescence 1:100-1:500, Immunohistochemistry-Paraffin 1:200-1:1000 |
| Applications | Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18699971
|
Novus Biologicals
NBP2-97164-100ul |
100 μg |
€ 326.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18600574
|
Novus Biologicals
NBP2-97164-50ul |
50 μg |
€ 227.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Aminopeptidase A/ENPEP Polyclonal antibody specifically detects Aminopeptidase A/ENPEP in Human samples. It is validated for Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
| Aminopeptidase A/ENPEP | |
| Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Adaptive Immunity, Cancer, Growth and Development, Hematopoietic Stem Cell Markers, Immunology, Neuronal Cell Markers, Stem Cell Markers | |
| PBS (pH 7.0) | |
| 2028 | |
| IgG | |
| Antigen and protein A Affinity-purified |
| Immunocytochemistry/ Immunofluorescence 1:100-1:500, Immunohistochemistry-Paraffin 1:200-1:1000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| Aminopeptidase A, APA, AP-A, CD249, CD249 antigen, Differentiation antigen gp160, EAP, EC 3.4.11, EC 3.4.11.7, glutamyl aminopeptidase, glutamyl aminopeptidase (aminopeptidase A), gp160 | |
| Produced in rabbits immunized with E. coli-derived Human Aminopeptidase A/ENPEP fragment. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title