missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aminopeptidase B/RNPEP Antibody, Janelia Fluor™ 549, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-18430JF549
This item is not returnable.
View return policy
Description
Aminopeptidase B/RNPEP Polyclonal antibody specifically detects Aminopeptidase B/RNPEP in Human samples. It is validated for Western Blot
Specifications
| Aminopeptidase B/RNPEP | |
| Polyclonal | |
| Western Blot | |
| aminopeptidase B, APB, AP-B, Arginine aminopeptidase, Arginyl aminopeptidase, arginyl aminopeptidase (aminopeptidase B), DKFZp547H084, EC 3.4.11, EC 3.4.11.6 | |
| Recognizes region between residues Between Between 600 and 650.. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| Janelia Fluor 549 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 6051 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction