missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aminopeptidase P1/XPNPEP1 Antibody (SAIC-07B-14), Alexa Fluor™ 350 , Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-20090AF350
This item is not returnable.
View return policy
Description
Aminopeptidase P1/XPNPEP1 Monoclonal antibody specifically detects Aminopeptidase P1/XPNPEP1 in Human samples. It is validated for Western Blot, ELISA
Specifications
| Aminopeptidase P1/XPNPEP1 | |
| Monoclonal | |
| Alexa Fluor 350 | |
| Aminoacylproline aminopeptidase, APP1, cytosolic, Cytosolic aminopeptidase P, EC 3.4.11.9, sAmp, xaa-Pro aminopeptidase 1, XPNPEPLX-prolyl aminopeptidase (aminopeptidase P)-like, X-prolyl aminopeptidase (aminopeptidase P) 1, soluble, X-prolyl aminopeptidase 1, soluble | |
| Peptide "e;GSLTFEPLTLVPIQTK"e; derived from Xaa-Pro aminopeptidase 1 conjugated to KLH. | |
| 0.1 mL | |
| metabolism, Signal Transduction | |
| 7511 | |
| Store at 4°C in the dark. | |
| IgG κ |
| Western Blot, ELISA | |
| SAIC-07B-14 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction