missing translation for 'onlineSavingsMsg'
Learn More

Aminopeptidase P2/XPNPEP2 Antibody (018), DyLight 594, Novus Biologicals™

Product Code. 18236286 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18236286 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18236286 Supplier Novus Biologicals Supplier No. NBP290014DL594

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

Aminopeptidase P2/XPNPEP2 Monoclonal specifically detects Aminopeptidase P2/XPNPEP2 in Human samples. It is validated for Sandwich ELISA Detection.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Aminopeptidase P2/XPNPEP2
Applications ELISA
Classification Monoclonal
Clone 18
Conjugate DyLight 594
Dilution Sandwich ELISA Detection
Formulation 50mM Sodium Borate
Gene Alias Aminoacylproline aminopeptidase, APP2, EC 3.4.11.9, mAmP, Membrane-bound aminopeptidase P, Membrane-bound AmP, Membrane-bound APP, xaa-Pro aminopeptidase 2, X-Pro aminopeptidase 2, X-prolyl aminopeptidase (aminopeptidase P) 2, membrane-bound
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human Aminopeptidase P2/XPNPEP2 (O43895; Met1-Ala650).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7512
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.