Learn More
AMMECR1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA562474
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: ASSSSHCSGE SQCRAGELGL GGAGTRLNGL GGLTGGGSGS GCTLSP Highest antigen sequence identity to the following orthologs: Mouse - 86%, Rat - 88%.
AMMECR1 Polyclonal antibody specifically detects AMMECR1 in Human samples. It is validated for Immunohistochemistry.Specifications
AMMECR1 | |
Polyclonal | |
Unconjugated | |
AMMECR1 | |
6230420G18Rik; Alport syndrome mental retardation midface hypoplasia and elliptocytosis chromosomal region protein 1; Alport syndrome, mental retardation, midface hypoplasia and elliptocytosis chromosomal region gene 1; Alport syndrome, mental retardation, midface hypoplasia and elliptocytosis chromosomal region gene 1 homolog; AMME syndrome candidate gene 1 protein; AMME syndrome candidate gene 1 protein homolog; Ammecr1; AMMERC1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
9949 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9Y4X0 | |
AMMECR1 | |
Recombinant protein corresponding to Human AMMECR1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only