missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Iron(III) Citrate, Reagent Grade, Honeywell™

Catalog Number p-7120180
Click to view available options
Quantity:
100 g
500 g
Packaging:
Poly bottle
This item is not returnable. View return policy
1185-57-5
C6H11FeNO7+3
264.999
FRHBOQMZUOWXQL-UHFFFAOYSA-N
ammonium ferric citrate, ferri seltz, ammonii ferri citras, ferric ammonium citrate, iron ammonium citrate, ammonium iron 3+ citrate, ammonium iron iii citrate, iron ammonium citrate, green, ferri-ammoniumcitrat, braunes, ferriseltz
118984355
azane;2-hydroxypropane-1,2,3-tricarboxylic acid;iron(3+)
C(C(=O)O)C(CC(=O)O)(C(=O)O)O.N.[Fe+3]
This item is not returnable. View return policy
1185-57-5
C6H11FeNO7+3
264.999
FRHBOQMZUOWXQL-UHFFFAOYSA-N
ammonium ferric citrate, ferri seltz, ammonii ferri citras, ferric ammonium citrate, iron ammonium citrate, ammonium iron 3+ citrate, ammonium iron iii citrate, iron ammonium citrate, green, ferri-ammoniumcitrat, braunes, ferriseltz
118984355
azane;2-hydroxypropane-1,2,3-tricarboxylic acid;iron(3+)
C(C(=O)O)C(CC(=O)O)(C(=O)O)O.N.[Fe+3]

Reagent Grade, powder

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY
Color Green
Physical Form Solid
Quantity 100 g
Linear Formula C6H8O7 ·xFe3+ · yNH3
Packaging Poly bottle
Chemical Name or Material Ammonium iron(III) citrate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'