missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Amnionless/AMN Antibody, DyLight 488, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97416G
This item is not returnable.
View return policy
Description
Amnionless/AMN Polyclonal specifically detects Amnionless/AMN in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| Amnionless/AMN | |
| Polyclonal | |
| Immunohistochemistry, Immunohistochemistry-Paraffin | |
| amnionless homolog (mouse), PRO1028, protein amnionless, UNQ513/PRO1028, visceral endoderm-specific type 1 transmembrane protein | |
| Produced in rabbits immunized with E. coli-derived Human Amnionless/AMN fragment. | |
| 0.1 mL | |
| metabolism | |
| 81693 | |
| Store at 4°C in the dark. | |
| IgG |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| DyLight 488 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction