missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AMOT Antibody (OTI1A8), Allophycocyanin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72429APC
This item is not returnable.
View return policy
Description
AMOT Monoclonal antibody specifically detects AMOT in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
Specifications
| AMOT | |
| Monoclonal | |
| APC | |
| PBS | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| 154796 | |
| Store at 4C in the dark. | |
| IgG1 |
| Western Blot, Immunocytochemistry | |
| OTI1A8 | |
| Western Blot, Immunocytochemistry/ Immunofluorescence | |
| angiomotin, angiomotin p80 isoform, KIAA1071angiomotin p130 isoform | |
| Full length human recombinant protein of human AMOT(NP_573572) produced in HEK293T cell. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction