missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AMOT Monoclonal Antibody (OTI3F9), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA507142
This item is not returnable.
View return policy
Description
AMOT (Angiomotin) belongs to the motin family of angiostatin binding proteins characterized by conserved coiled-coil domains and C-terminal PDZ binding motifs. AMOT plays a central role in tight junction maintece via the complex formed with ARHGAP17 - which acts by regulating the uptake of polarity proteins at tight junctions. AMOT is expressed predomitly in endothelial cells of capillaries as well as larger vessels of the placenta where it may mediate the inhibitory effect of angiostatin on tube formation and the migration of endothelial cells toward growth factors during the formation of new blood vessels.Specifications
| AMOT | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q4VCS5 | |
| AMOT | |
| Full length human recombit protein of human AMOT produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI3F9 | |
| Unconjugated | |
| AMOT | |
| angiomotin | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 154796 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction