Description
Honeywell Fluka™ offers an expanding range of high quality inorganic certified reference materials and organic reference materials.
- Produced according to ISO 9001
- Suitable for a wide range of industries and applications
- Detailed Certificate of Analysis included with every delivery
- Expert Technical Support
Specifications
Specifications
| Chemical Name or Material | AMOZ-d5 |
| Packaging | Glass Bottle |
| CAS | 1017793-94-0 |
| Color | White |
| Melting Point | 118°C to 120°C |
| Synonym | 3-Amino-5-morpholinomethyl-2-oxazolidinone-d5, 4,4,5,a,a-Pentadeutero-3-amino-5-morpholinomethyl-2-oxazolidinone |
| Molecular Weight (g/mol) | 206.22 |
| Quantity | 10 mg |
| Grade | Analytical Standard |