Learn More
AMOZ-d5, TRC
Specifications
Specifications
| Chemical Name or Material | 3-Amino-5-morpholinomethyl-2-oxazolidinone D5 (AMOZ D5) |
| CAS | 1017793-94-0 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C8 2H5 H10 N3 O3 |
| Quantity | 10 mg |
| Synonym | 3-Amino-5-morpholinomethyl-2-oxazolidinone D5 (AMOZ D5), 2-Oxazolidinone-4,4,5-d3, 3-amino-5-(4-morpholinylmethyl-d2)- |
| InChI Formula | InChI=1S/C8H15N3O3/c9-11-6-7(14-8(11)12)5-10-1-3-13-4-2-10/h7H,1-6,9H2/i5D2,6D2,7D |
| SMILES | [2H]C([2H])(N1CCOCC1)C2([2H])OC(=O)N(N)C2([2H])[2H] |
| IUPAC Name | 3-amino-4,4,5-trideuterio-5-[dideuterio(morpholin-4-yl)methyl]-1,3-oxazolidin-2-one |
| Molecular Weight (g/mol) | 206.25 |
| Show More |