missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ AMPD2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA583791
This item is not returnable.
View return policy
Description
Immunogen sequence: MDGKCKEIAE ELFTRSLAES ELRSAPYEFP EESPIEQLEE RRQRLERQIS QDVKLEPDIL LRAKQDFLKT DSDSDLQLYK EQGE Highest antigen sequence indentity to the following orthologs: Rat - 99%, Mouse - 99%.
Adenosine monophosphate deaminase-2 (EC 3.5.4.6) catalyzes the deamination of AMP to IMP and plays an important role in the purine nucleotide cycle.
Specifications
| AMPD2 | |
| Polyclonal | |
| Unconjugated | |
| Ampd2 | |
| 1200014F01Rik; adenosine monophosphate deaminase 2; adenosine monophosphate deaminase 2 (isoform L); adenosine monophosphate deaminase 2 tv2; AI552571; AMP deaminase 2; AMP deaminase isoform L; AMPD; AMPD2; Ampd-2; PCH9; SPG63 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 271 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q01433 | |
| Ampd2 | |
| Recombinant protein corresponding to Human AMPD2. Recombinant protein control fragment (Product #RP-94074). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction